Order Entry
United Kingdom
ContactUsLinkComponent
Anti-Rabbit IgG H&L Goat Polyclonal Antibody
Anti-Rabbit IgG H&L Goat Polyclonal Antibody
Catalog # ANTIA294873-500
Supplier:  ANTIBODIES.COM
New Product
Anti-Rabbit IgG H&L Goat Polyclonal Antibody
Catalog # ANTIA294873-500
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Antibody type:
    Secondary
  • Antigen symbol:
    Rabbit IgG H&L
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Goat
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Rabbit
  • Western blot:
    Yes
  • Form:
    Liquid
  • Antibody fragment:
    Whole Molecule
  • Storage buffer:
    Supplied in phosphate buffered saline with 0,09% sodium azide
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    0,5 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Tested applications:
    IHC, ICC
  • Purification:
    Antigen affinity purification
  • Pack type:
    Plastic vial
  • Pk:
    500 µG

Specifications

About this item

Goat anti-rabbit IgG H&L secondary antibody for WB, IHC, ICC and ELISA with samples derived from rabbit.

  • Validated applications: WB, IHC, ICC, ELISA
  • Recommended dilutions: WB: 1:1000 to 1:20000, IHC: 1:100 to 1:1000, ICC: 1:100 to 1:1000, ELISA: 1:1000 to 1:20000; ELISA (capture antibody): 1:50 to 1:250

Type: Secondary
Antigen: Rabbit IgG H&L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rabbit