Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Mouse Thrombomodulin ELISA Kit

Mouse Thrombomodulin ELISA Kit

Mouse Thrombomodulin ELISA kit, 96 Tests
Catalog # ANTIA311191-96
Supplier:   Antibodies.com
Mouse Thrombomodulin ELISA Kit
Catalog # ANTIA311191-96
Supplier:  Antibodies.com
CAS Number:  
Mouse Thrombomodulin ELISA Kit
Mouse Thrombomodulin ELISA Kit

Mouse Thrombomodulin ELISA kit, 96 Tests

CAS Number:  
Price
£450.00
Log in for availability
1 * 96 Tests
Product Details & Documents

Mouse Thrombomodulin ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse Thrombomodulin in serum, plasma, and other biological fluids.

  • Higher throughput: Get results in just 90 minutes, with a single wash step
  • Detection range: 7,5 to 120 ng/ml
  • Sensitivity: 0,23 ng/ml
  • Sample type: Serum, plasma or other biological fluids
  • Assay precision: Intra assay: CV <8%, Inter assay: CV <10%

Mouse Thrombomodulin ELISA kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse Thrombomodulin in An antibody specific for Thrombomodulin has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Thrombomodulin present in each sample is bound to the wells by the immobilised antibody. Biotinylated Anti-Thrombomodulin antibody, which also binds the Thrombomodulin present in each sample, and Streptavidin-HRP, which binds the Biotinylated Anti-Thrombomodulin antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated Anti-Thrombomodulin antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. colour develops in proportion to the amount of Thrombomodulin captured in each well. The colour development is stopped by addition of stop solution which changes the colour from blue to yellow and the intensity of the colour is then measured. The concentration of Thrombomodulin in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, standard solution, standard diluent, biotinylated detection antibody, streptavidin-HRP, wash buffer (25X), substrate solution A, substrate solution B, stop solution, plate sealers, foil pouch, and protocol manual.

Specifications

Specifications

  • Pk:
    96 Tests
  • Description:
    Mouse Thrombomodulin ELISA kit
  • Primary antibody reactivity:
    Mouse
  • Sample type:
    Serum, plasma or other biological fluids
  • Sample volume:
    40 μl
  • Detection range:
    7,5 - 120 ng/ml
  • Storage temperature:
    4 °C
  • Shelf life:
    Store for 6 months at 4 °C
  • Assay Type:
    Sandwich
  • Time to Results:
    1 h 30 min
  • Cat. No.:
    ANTIA311191-96
  • Assay duration:
    One step
  • Detection method:
    Colorimetric
  • Sensitivity:
    0,23 ng/ml
  • Format:
    Pre-coated
  • Target protein:
    Thrombomodulin
  • Regulatory status:
    RUO

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.