Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Human GCP2 ELISA Kit

Human GCP2 ELISA Kit

Human GCP2 ELISA kit, 96 Tests
Catalog # ANTIA311881-96
Supplier:   Antibodies.com
Human GCP2 ELISA Kit
Catalog # ANTIA311881-96
Supplier:  Antibodies.com
CAS Number:  
Human GCP2 ELISA Kit
Human GCP2 ELISA Kit

Human GCP2 ELISA kit, 96 Tests

CAS Number:  
Price
£450.00
Log in for availability
1 * 96 Tests
Product Details & Documents

Human GCP2 ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human GCP2 in serum, plasma, and other biological fluids.

  • Higher throughput: Get results in just 90 minutess, with a single wash step
  • Detection range: 0,4 to 6,4 ng/ml
  • Sensitivity: 0,022 ng/ml
  • Sample Type: Serum, plasma or other biological fluids
  • Assay precision: Intra assay: CV <8%, Inter assay: CV <10%

Human GCP2 ELISA kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of human GCP2 in serum, plasma or other biological fluids. An antibody specific for GCP2 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the GCP2 present in each sample is bound to the wells by the immobilised antibody. Biotinylated anti-GCP2 antibody, which also binds the GCP2 present in each sample, and Streptavidin-HRP, which binds the Biotinylated anti-GCP2 antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated anti-GCP2 antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. colour develops in proportion to the amount of GCP2 captured in each well. The colour development is stopped by addition of stop solution which changes the colour from blue to yellow and the intensity of the colour is then measured. The concentration of GCP2 in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, standard solution, standard diluent, biotinylated detection antibody, steptavidin-HRP, wash buffer (25X), substrate solution A, substrate solution B, stop solution, plate sealers, foil pouch, and protocol manual.

Specifications

Specifications

  • Pk:
    96 Tests
  • Description:
    Human GCP2 ELISA kit
  • Primary antibody reactivity:
    Human
  • Sample type:
    Serum, plasma or other biological fluids
  • Sample volume:
    40 μl
  • Detection range:
    0,4 - 6,4 ng/ml
  • Storage temperature:
    4 °C
  • Shelf life:
    Store for 6 months at 4 °C
  • Assay Type:
    Sandwich
  • Time to Results:
    1 h 30 min
  • Cat. No.:
    ANTIA311881-96
  • Assay duration:
    One step
  • Detection method:
    Colorimetric
  • Sensitivity:
    0,022 ng/ml
  • Format:
    Pre-coated
  • Target protein:
    GCP2
  • Regulatory status:
    RUO

Specifications