Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Rat WISP1 ELISA Kit

Rat WISP1 ELISA Kit

Rat WISP1 ELISA kit, 96 Tests
Catalog # ANTIA79796-96
Supplier:   Antibodies.com
Rat WISP1 ELISA Kit
Catalog # ANTIA79796-96
Supplier:  Antibodies.com
CAS Number:  
Rat WISP1 ELISA Kit
Rat WISP1 ELISA Kit

Rat WISP1 ELISA kit, 96 Tests

CAS Number:  
Price
£460.00
Log in for availability
1 * 96 Tests
Product Details & Documents

Rat WISP1 ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat WISP1 in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready-To-Use ELISA Kit
  • Detection Range: 15,625 - 1000 pg/ml
  • Sensitivity: 9,375 pg/ml
  • Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay Precision: Intra-Assay: CV <8%, Inter-Assay: CV <10%

Rat WISP1 ELISA kit (A79796) employs the sandwich enzyme immunoassay technique for the quantitative measurement of rat WISP1 in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for WISP1 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the WISP1 present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-WISP1 Antibody, which binds the captured WISP1 present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of WISP1 captured in each well. The concentration of WISP1 can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

 : This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin Conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.

Specifications

Specifications

  • Pk:
    96 Tests
  • Description:
    Rat WISP1 ELISA kit
  • Primary antibody reactivity:
    Rat
  • Sample type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Sample volume:
    100 μl
  • Detection range:
    15,625 - 1000 pg/ml
  • Storage temperature:
    4 °C
  • Shelf life:
    Store for 6 months at 4 °C
  • Assay Type:
    Sandwich
  • Time to Results:
    4 h 30 min
  • Cross reactivity:
    Rat WISP1 ELISA kit exhibits high specificity and excellent specificity for the detection of rat WISP1. No significant cross-reactivity or interference between WISP1 and analogues was observed.
  • Assay Principle:
    Quantitative
  • Cat. No.:
    ANTIA79796-96
  • Assay duration:
    Multiple steps
  • Detection method:
    Colorimetric
  • Sensitivity:
    9,375 pg/ml
  • Format:
    Pre-coated
  • Target protein:
    WISP1
  • Regulatory status:
    RUO

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.