Order Entry
United Kingdom
ContactUsLinkComponent
Idex® Inline Chromatography Check Valves
Idex® Inline Chromatography Check Valves
Supplier:  Cole-Parmer
New Product
Certificates
Idex® Inline Chromatography Check Valves
Supplier:  Cole-Parmer
New Product
Specifications
  • Catalog No:
  • COLE02013-34
  • COLE02013-35
  • Max. pressure:
  • 138 bar
  • 138 bar
  • Orifice:
  • 0.02
  • 0.02
  • Wetted materials:
  • PEEK/perfluoroelastomer/SS
  • PEEK/perfluoroelastomer/SS
  • Warranty:
  • 1 year
  • 1 year