Order Entry
United Kingdom
ContactUsLinkComponent
n-Butyl phenyl ether
Irritant
n-Butyl phenyl ether
Catalog # ACSDSB68178.1G
Supplier:  CPACHEM
New Product
CAS Number:  
Irritant
n-Butyl phenyl ether
Catalog # ACSDSB68178.1G
Supplier:  CPACHEM
New Product
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :150.22 g/mol
 :GHS07
CAS Number:1126-79-0
Formula:C₁₀H₁₄O
MDL Number:MFCD00009438