About this item
Rat monoclonal IgG1 antibody against human, mouse, dog E-cadherin (CD324), unconjugated.
- Airway cells
The DECMA-1 antibody recognizes E-cadherin, a calcium-dependent transmembrane glycoprotein involved in cell-cell adhesion, bacterial pathogenesis, and tumor invasion. The extracellular region of E-cadherin is composed of multiple cadherin repeats that contain calcium-binding motifs. The cytoplasmic domain of E-cadherin is reported to interact with a range of intracellular proteins including erbin, ezrin, caspase-3, caspase-8, β-catenin, presenilin 1, and casein kinase II. E-cadherin is expressed in most epithelial cells and in erythroid cells. The E-cadherin antibody is often used as a neutralizing antibody to inhibit E-cadherin-mediated cell-cell interactions.
Type:
Antigen: E-cadherin (CD324)
Clonality: Monoclonal
Clone: DECMA-1
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1 kappa
Reactivity: Dog;Human;Mouse;Pig
Specifications
- Catalog No:
- STMC60157.1
- STMC60157
- Antigen name:
- E-cadherin (CD324)
- E-cadherin (CD324)
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- DECMA-1
- DECMA-1
- Reactivity:
- Pig, Human, Dog, Mouse
- Pig, Human, Dog, Mouse
- Host:
- Rat
- Rat
- Gene ID:
- 999/12550
- 999/12550
- Isotype:
- IgG1 kappa
- IgG1 kappa