- Antigen name:Beta-tubulin III
- Clonality:Monoclonal
- Clone:TUJ1
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Other,Human,Rat,Mouse
- Gene ID:10381
- Pk:250 µl
Mouse monoclonal IgG2a antibody against human, mouse, rat beta-tubulin III, unconjugated.
- Neural cells Psc derived
- Neurons
The TUJ1 antibody reacts with beta-tubulin III, an ~50 to 55 kDa structural protein that is a component of tubulin. Tubulin is the major component of microtubules within the cytoskeleton and is assembled from heterodimers of alpha and beta tubulin subunits. The beta III isoform of tubulin, also known as neuron-specific class III beta-tubulin, is expressed primarily in neurons and is widely used as a marker to distinguish neurons from other cell types. Beta-tubulin III contributes to microtubule formation in neuronal cell bodies and axons, a function involving GTP binding, and plays roles in axonal transport, neuronal cell proliferation, and differentiation. It is highly expressed in several types of cancer and is a predictive and prognostic marker for various tumors, for example, being found in neoplastic but not in normal glial cells. The TUJ1 antibody is expected to recognise all mammalian homologs of beta-tubulin III and the epitope has reportedly been mapped to the C-terminal 15 amino acids of the protein.
Type:
Antigen: Beta-tubulin III
Clonality: Monoclonal
Clone: TUJ1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human, Mouse, Other, Rat