Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
 

Human Recombinant Myostatin

Marketsource
Human Recombinant Myostatin
Marketsource
Certificates
Human Recombinant Myostatin

2 Options Available Below

Product Details & Documents

Myostatin is a member of transforming growth factor beta (TGF-β) superfamily that signals by binding to activin type II receptor, resulting in the recruitment of either ALK3 or ALK4 coreceptor, and activation of SMAD2/3 (Kondás et al.). It is released by myocytes and inhibits muscle growth and ability to regenerate (Lee and Lee). Myostatin appears to affect the lipid catabolic metabolism of adipocytes and inhibits preadipocyte differentiation, as demonstrated in the 3T3-L1 cell line (Li et al.).

 : Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • STMC78119
  • STMC78119.1
  • Protein/peptide name:
  • Myostatin
  • Myostatin
  • Protein/peptide type:
  • Recombinant
  • Recombinant
  • Species:
  • Human
  • Human
  • Purity:
  • ≥95%
  • ≥95%

Specifications

Product Family Options

Product Information

  • Pk
    Availability
    Price
  • 10 µG
    Log in for availability
    Log in for availability
    1 * 10 µG
    1 * 10 µG - £159.00
    £159.00
    1 * 10 µG
       £159.00

    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    STMC78119
    Protein/peptide name
    Myostatin
    Protein/peptide type
    Recombinant
    Species
    Human
    Purity
    ≥95%
    Marketsource
  • 100 µG
    Log in for availability
    Log in for availability
    1 * 100 µG
    1 * 100 µG - £834.00
    £834.00
    1 * 100 µG
       £834.00

    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    STMC78119.1
    Protein/peptide name
    Myostatin
    Protein/peptide type
    Recombinant
    Species
    Human
    Purity
    ≥95%
    Marketsource