Kinesis KX Non Sterile Syringe Filter, PVDF
Supplier: Cole-Parmer
New Product
Certificates
About this item
Resistant to many solvents and show low protein binding - ideal for general filtration of biological samples.
- Broad chemical compatibility and low UV absorbing extractable
- Solvent-resistant filter housing with nominal extractable
- Leak-free Luer-Lok® and Luer-slip connections
- Color-coded filters for easy organization
Prepare reliable aqueous, organic, and biomolecular samples with membrane filtration inside a polypropylene (PP) housing.
Syringe filters have Luer-Lok and Luer-slip connections for easy, quick and tight connections to tubing, syringes, or needles. Filter packs are available in a resealable storage container to prevent contamination during multiple opening and closings.
Specifications
- Catalog No:
- COLE12917-09
- COLE12917-12
- Max. operating pressure:
- 100 psi
- 100 psi
Specifications
Frequently Bought Together