Order Entry
United Kingdom
ContactUsLinkComponent
meta-Phosphoric acid ≥33,5% BioXtra
Corrosive
meta-Phosphoric acid ≥33,5% BioXtra
Catalog # SIALM6288-100G
Supplier:  Merck
New Product
CAS Number:  
Corrosive
meta-Phosphoric acid ≥33,5% BioXtra
Catalog # SIALM6288-100G
Supplier:  Merck
New Product
CAS Number:  

Specifications

  • Pk:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • Appearance (Colour):
    Colourless or White
  • Appearance (Form):
    Powder or Crystals
  • Insoluble matter:
    ≤0.1%
  • Ammonia (NH4):
    ≤0.05%
  • Assay:
    ≥33.5%
  • Chloride Content:
    ≤0.001%
  • Sulfate:
    ≤0.005%
  • Stabilizer:
    ≥57.0%
  • Aluminum (AL):
    ≤0.0005%
  • Calcium (Ca):
    ≤0.005%
  • Copper (Cu):
    ≤0.0005%
  • Iron (Fe):
    ≤0.0005%
  • Lead (Pb):
    ≤0.001%
  • Magnesium (Mg):
    ≤0.001%
  • Zinc (Zn):
    ≤0.0005%
  • Potassium (K):
    ≤0.005%

Specification Test Results

About this item

 :
 :79.98 g/mol
Boiling Pt:260 °C (1013 hPa)
Melting Pt:200 °C
Density:2 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:253-433-4
UN:3260
ADR:8,III
 :M-Clarity™ quality level = MQ200
 :GHS05
CAS Number:37267-86-0
Formula:(HPO₃)n
MDL Number:MFCD00011351