Anti-NUDT9 Rabbit Polyclonal Antibody
Catalog # PRSI56-294
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:ADP-ribose pyrophosphatase, mitochondrial
- Antigen symbol:NUDT9
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Western blot:Yes
- Epitope:Amino Acid Range = 318-346
- Form:Liquid
- Gene ID:Q9BW91
- Antigen synonyms:NUDT9
- Modification:None
- Storage buffer:Supplied in PBS with 0,09% (W/V) sodium azide
- Molecular weight:39 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,32 mg/ml
- Shipping temperature:4 °C
- Immunogen:This NUDT9 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 318-346 amino acids from the C-terminal region of human NUDT9.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Size:400 µl
- Pk:400 µl
Specifications
About this item
NUDT9 hydrolyzes ADP-ribose (ADPR) to AMP and ribose 5'-phosphate.
For WB starting dilution is: 1:1000. For IHC-P starting dilution is: 1:10~50.
Type: Primary
Antigen: NUDT9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Human, Mouse