Anti-POLR3GL Rabbit Polyclonal Antibody
PRSI55-809
New Product
- Antibody type:Primary
- Antigen name:DNA-directed RNA polymerase III subunit RPC7-like
- Antigen symbol:POLR3GL
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Mouse
- Western blot:Yes
- Epitope:Amino Acid Range = 65-93
- Form:Liquid
- Gene ID:Q9BT43
- Antigen synonyms:POLR3GL
- Modification:None
- Storage buffer:Supplied in PBS with 0,09% (W/V) sodium azide
- Molecular weight:25 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,5 mg/ml
- Shipping temperature:4 °C
- Immunogen:This POLR3GL antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 65-93 amino acids from the Central region of human POLR3GL.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Size:400 µl
- Pk:400 µl
For WB starting dilution is: 1:1000 For IHC-P starting dilution is: 1:50~100
Type: Primary
Antigen: POLR3GL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Mouse