Anti-HIP1R Rabbit Polyclonal Antibody
Catalog # PRSI55-634
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:Huntingtin-interacting protein 1-related protein
- Antigen symbol:HIP1R
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:Amino Acid Range = 98-126
- Form:Liquid
- Gene ID:O75146
- Antigen synonyms:HIP1R
- Modification:None
- Storage buffer:Supplied in PBS with 0,09% (W/V) sodium azide
- Molecular weight:119 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,32 mg/ml
- Shipping temperature:4 °C
- Immunogen:This HIP1R antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 98-126 amino acids from the N-terminal region of human HIP1R.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Size:400 µl
- Pk:400 µl
Specifications
About this item
This protein is a "fusion" protein encoding four enzymatic activities of the pyrimidine pathway (GATase, CPSase, ATCase and DHOase).
For WB starting dilution is: 1:1000. For IHC-P starting dilution is: 1:10~50. For FACS starting dilution is: 1:10~50.
Type: Primary
Antigen: HIP1R
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Human