Anti-IFT81 Rabbit Polyclonal Antibody
Catalog # PRSI55-234
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:Intraflagellar transport protein 81 homolog
- Antigen symbol:IFT81
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:Amino Acid Range = 527-555
- Form:Liquid
- Gene ID:Q8WYA0
- Antigen synonyms:IFT81
- Modification:None
- Storage buffer:Supplied in PBS with 0,09% (W/V) sodium azide
- Molecular weight:80 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,5 mg/ml
- Shipping temperature:4 °C
- Immunogen:This IFT81 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 527-555 amino acids from the C-terminal region of human IFT81.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Size:400 µl
- Pk:400 µl
Specifications
About this item
Isoform CDV-1 may be involved in cardiac hypertrophy caused by carnitine deficiency (By similarity). Isoform CDV-1R appears to play a role in development of the testis and spermatogenesis (By similarity).
For WB starting dilution is: 1:1000.
Type: Primary
Antigen: IFT81
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Human, Mouse