Anti-NIP Recombinant Antibody [Clone: B1-8]
PRSI24-090
- Antibody type:Primary
- Clonality:Recombinant
- Clone:B1-8
- Conjugation:Unconjugated
- Isotype:Fab fragment lambda
- Cross adsorption:No
- Form:Liquid
- Antigen synonyms:NIP
- Modification:Unmodified
- Storage buffer:PBS with 0,02% Proclin 300
- Storage temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping temperature:Shipped on blue ice
- Immunogen:hapten-protein conjugated NP12-chicken gammaglobulin (CG).
- Tested applications:negative control
- Size:0,2 mg
- Pack type:Vial
- Pk:0,2 mg
Type: Primary
Antigen: NIP
Clonality: Recombinant
Clone: B1-8
Conjugation: Unconjugated
Host: Recombinant
Isotype: Fab fragment lambda
Reactivity: