Anti-MYC Recombinant Antibody [Clone: 9E10]
PRSI24-046
New Product
- Antibody type:Primary
- Antigen name:MYC proto-oncogene, bHLH transcription factor
- Antigen symbol:MYC
- Clonality:Recombinant
- Clone:9E10
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2a kappa
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Gene ID:4609
- Antigen synonyms:cmyc|Class E basic helix-loop-helix protein 39|Proto-oncogene c-Myc|BHLHE39|Transcription factor p64
- Modification:Unmodified
- Storage buffer:PBS with 0,02% Proclin 300
- Storage temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping temperature:Shipped on blue ice
- Immunogen:A synthetic peptide corresponding to aa 408-439 from C-terminus of human c-myc.
- Size:0,2 mg
- Pack type:Vial
- Pk:0,2 mg
Type: Primary
Antigen: MYC proto-oncogene, bHLH transcription factor
Clonality: Recombinant
Clone: 9E10
Conjugation: Unconjugated
Host: Recombinant
Isotype: IgG2a kappa
Reactivity: Human