Anti-EDAR Rabbit Polyclonal Antibody
Catalog # PRSI15-374
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:Ectodysplasin A receptor
- Antigen symbol:EDAR
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:10913
- Antigen synonyms:EDAR|ECTD10B|ED5|ED3|ED1R|HRM1|EDA-A1R|ECTD10A|EDA1R|EDA3|DL
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 27-187 of human EDAR (NP_071731.1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: EDAR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat