Anti-ILF3 Rabbit Polyclonal Antibody
PRSI15-318
New Product
- Antibody type:Primary
- Antigen name:Interleukin enhancer binding factor 3, 90 kDa
- Antigen symbol:ILF3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:3609
- Antigen synonyms:MPHOSPH4|NFAR2|MMP4|TCP80|DRBF|NF90a|CBTF|MPP4|NF90|NF-AT-90|DRBP76|NF90b|NF110|ILF3|NFAR-1|NF110b|NFAR|TCP110
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 597-706 of human ILF3 (NP_001131145.1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: ILF3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat