Anti-RASEF Rabbit Polyclonal Antibody
Catalog # PRSI15-247
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:RAS and EF-hand domain containing
- Antigen symbol:RASEF
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:158158
- Antigen synonyms:ras-related protein Rab-45|ras and EF-hand domain-containing protein|RAB45
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 200-500 of human RASEF (NP_689786.2)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: RASEF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat