Order Entry
United Kingdom
ContactUsLinkComponent
Anti-CHST11 Rabbit Polyclonal Antibody
Anti-CHST11 Rabbit Polyclonal Antibody
Catalog # PRSI15-172
Supplier:  ProSci Inc.
New Product
Anti-CHST11 Rabbit Polyclonal Antibody
Catalog # PRSI15-172
Supplier:  ProSci Inc.
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Carbohydrate sulfotransferase 11
  • Antigen symbol:
    CHST11
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    50515
  • Antigen synonyms:
    HSA269537|C4ST|carbohydrate (chondroitin 4) sulfotransferase 11|C4ST1|C4S-1|chondroitin 4-O-sulfotransferase 1|C4ST-1|IgH/CHST11 fusion|carbohydrate sulfotransferase 11|OCBMD
  • Modification:
    None
  • Storage buffer:
    PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
  • Storage temperature:
    Store at −20 °C. Avoid freeze/thaw cycles
  • Concentration:
    Batch dependent
  • Shipping temperature:
    20
  • Immunogen:
    Recombinant fusion protein containing a sequence corresponding to amino acids 38-352 of human CHST11 (NP_060883.1)
  • Purification:
    Affinity purification
  • Pack type:
    Vial
  • Pk:
    50 µl

Specifications

About this item

Type: Primary
Antigen: CHST11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat