Anti-NAT8B Rabbit Polyclonal Antibody
Catalog # PRSI14-984
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:N-acetyltransferase 8B (putative, gene/pseudogene)
- Antigen symbol:NAT8B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Form:Liquid
- Gene ID:51471
- Antigen synonyms:N-acetyltransferase 8B (GCN5-related|ATase1|probable N-acetyltransferase 8B|NAT8B|Hcml2P|putative N-acetyltransferase 8B|gene/pseudogene)|putative|acetyltransferase 1|CML2|N-acetyltransferase Camello 2|camello-like protein 2
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 75-165 of human NAT8B (NP_057431.2)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: NAT8B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human