Anti-DPP3 Rabbit Polyclonal Antibody
Catalog # PRSI14-966
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:Dipeptidyl peptidase 3
- Antigen symbol:DPP3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:10072
- Antigen synonyms:Dipeptidyl arylamidase III|DPP III|Enkephalinase B|Dipeptidyl peptidase 3|Dipeptidyl aminopeptidase III|Dipeptidyl peptidase III|DPP3
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-280 of human DPP3 (NP_569710.2)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: DPP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat