Anti-TEAD1 Rabbit Polyclonal Antibody
PRSI14-950
New Product
- Antibody type:Primary
- Antigen name:TEA domain family member 1 (SV40 transcriptional enhancer factor)
- Antigen symbol:TEAD1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- ChIP:Yes
- Form:Liquid
- Gene ID:7003
- Antigen synonyms:TCF-13|NTEF-1|AA|TCF13|TEF-1|REF1|TEAD-1|TEAD1
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 135-215 of human TEAD1 (NP_068780.2)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: TEAD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse