Anti-Gabrb2 Rabbit Monoclonal Antibody [clone: H3]
PRSI54-329
New Product
- Antibody type:Primary
- Antigen name:gamma-aminobutyric acid (GABA) A receptor, subunit beta 2
- Antigen symbol:Gabrb2
- Clonality:Monoclonal
- Clone:H3
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Western blot:Yes
- Gene ID:14401
- Antigen synonyms:AI834970|C030021G16Rik|C030002O17Rik|Gabrab2|Gabrb-2
- Modification:None
- Storage buffer:PBS, pH 7,2, 0,02% NaN₃
- Storage temperature:Store at 4 °C for up to 6 months. Store at −20 °C long term
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Synthetic peptide derived from mouse Gabrb2 (CEKAANANNEKMRLD)
- Purification:Protein A purified
- Pack type:Vial
- Pk:100 µG
Type: Primary
Antigen: Gabrb2
Clonality: Monoclonal
Clone: H3
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse