Anti-NUDT1 Rabbit Polyclonal Antibody
Catalog # PRSI14-924
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:nudix hydrolase 1
- Antigen symbol:NUDT1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:4521
- Antigen synonyms:8-oxo-dGTPase|2-hydroxy-dATP diphosphatase|nucleoside diphosphate-linked moiety X motif 1|8-dihydroguanosine triphosphatase|8-dihydro-8-oxoguanine triphosphatase|8-oxo-7|8-dihydrodeoxyguanosine triphosphatase|7|nucleoside diphosphate-linked moiety X-type motif 1|mutT human homolog 1|nudix motif 1|MTH1|nudix (nucleoside diphosphate linked moiety X)-type motif 1
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at –20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-179 of human NUDT1 (NP_945192.1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: NUDT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human