Anti-LIMS2 Rabbit Polyclonal Antibody
PRSI14-859
New Product
- Antibody type:Primary
- Antigen name:LIM and senescent cell antigen-like domains 2
- Antigen symbol:LIMS2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat
- Western blot:Yes
- Form:Liquid
- Gene ID:55679
- Antigen synonyms:PINCH2|LIM and senescent cell antigen-like domains 2|FLJ10044|OTTHUMP00000203848|LIMS2|ILK-binding protein|OTTHUMP00000203850|OTTHUMP00000203866
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at –20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-341 of human LIMS2 (NP_001154875.1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: LIMS2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat