Anti-SLC5A11 Rabbit Polyclonal Antibody
Catalog # PRSI14-829
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:solute carrier family 5 (sodium/inositol cotransporter), member 11
- Antigen symbol:SLC5A11
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:115584
- Antigen synonyms:KST1|RKST1|SMIT2|SLC5A11|SGLT6
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at –20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 540-640 of human SLC5A11 (NP_443176.2)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: SLC5A11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat