Anti-PITPNM1 Rabbit Polyclonal Antibody
Catalog # PRSI14-773
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:phosphatidylinositol transfer protein, membrane-associated 1
- Antigen symbol:PITPNM1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:9600
- Antigen synonyms:phosphatidylinositol transfer protein|PITPNM|NIR2|DRES9|PYK2 N-terminal domain-interacting receptor 2|Drosophila retinal degeneration B|membrane-associated
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at –20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 110-190 of human PITPNM1 (NP_004901.2)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: PITPNM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat