Anti-ITGB1BP1 Rabbit Polyclonal Antibody
Catalog # PRSI14-772
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:integrin beta 1 binding protein 1
- Antigen symbol:ITGB1BP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:9270
- Antigen synonyms:ICAP-1B|ICAP1|ITGB1BP1|ICAP-1alpha|ICAP-1|ICAP1A|ICAP1B|ICAP-1A|Bodenin
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at –20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human ITGB1BP1 (NP_004754.1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: ITGB1BP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human