Anti-NDUFS8 Rabbit Polyclonal Antibody
Catalog # PRSI14-757
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:NADH dehydrogenase (ubiquinone) Fe-S protein 8, 23kDa (NADH-coenzyme Q reductase)
- Antigen symbol:NDUFS8
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:4728
- Antigen synonyms:23kDa (NADH-coenzyme Q reductase)|CI23KD|NADH dehydrogenase (ubiquinone) Fe-S protein 8|NDUFS8|CI-23k|NADH-ubiquinone oxidoreductase 23 kDa subunit|NADH dehydrogenase ubiquinone Fe-S 8|complex I-23kD|TYKY
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at –20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 35-210 of human NDUFS8 (NP_002487.1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: NDUFS8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat