Anti-IFI30 Rabbit Polyclonal Antibody
Catalog # PRSI14-734
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:IFI30 lysosomal thiol reductase
- Antigen symbol:IFI30
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:10437
- Antigen synonyms:interferon|GILT|gamma-inducible protein 3legumaturain|gamma-interferon-inducible protein IP-3interferon gamma-inducible protein 30 preproprotein|IFI-3IP-3IP3gamma-interferon-inducible lysosomal thiol reductase
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at –20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-250 of human IFI30 (NP_006323.2)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: IFI30
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human