Anti-PITPNA Rabbit Polyclonal Antibody
PRSI14-728
New Product
- Antibody type:Primary
- Antigen name:phosphatidylinositol transfer protein, alpha
- Antigen symbol:PITPNA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:5306
- Antigen synonyms:phosphotidylinositol transfer protein|phosphatidylinositol transfer protein|PITPNB|PITPN|PITP alpha
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at –20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 211-270 of human PITPNA (NP_006215.1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: PITPNA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat