Order Entry
United Kingdom
ContactUsLinkComponent
Anti-EP400 Rabbit Polyclonal Antibody
Anti-EP400 Rabbit Polyclonal Antibody
Catalog # PRSI14-281
Supplier:  ProSci Inc.
Anti-EP400 Rabbit Polyclonal Antibody
Catalog # PRSI14-281
Supplier:  ProSci Inc.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    E1A binding protein p400
  • Antigen symbol:
    EP400
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    57634
  • Antigen synonyms:
    hDomino|CAGH32|P40TNRC12|trinucleotide repeat containing 12|p400 kDa SWI2/SNF2-related protein|p400 SWI2/SNF2-related protein|domino homolog|trinucleotide repeat-containing gene 12 protein|hDomino(p400)|E1A-binding protein p40CAG repeat protein 32
  • Modification:
    None
  • Storage buffer:
    PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
  • Storage temperature:
    Store at –20...+730 °C. Avoid freeze/thaw cycles.
  • Concentration:
    Batch dependent
  • Shipping temperature:
    20
  • Immunogen:
    Recombinant fusion protein containing a sequence corresponding to amino acids 90-330 of human EP400 (NP_056224.3)
  • Purification:
    Affinity purification
  • Pack type:
    Vial
  • Pk:
    50 µl

Specifications

About this item

Type: Primary
Antigen: EP400
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human