Anti-CAMK2G Rabbit Polyclonal Antibody
Catalog # PRSI62-733
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:Calcium/calmodulin-dependent protein kinase type II subunit gamma
- Antigen symbol:CAMK2G
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:309-338 aa
- Form:Liquid
- Gene ID:818
- Modification:None
- Molecular weight:63 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:2 mg/ml
- Shipping temperature:4 °C
- Immunogen:This CAMK2G (CAMK2 gamma) antibody is generated from rabbits immunised with a KLH conjugated synthetic peptide between 309-338 amino acids from the C-terminal region of human CAMK2G (CAMK2 gamma).
- Purification:This antibody is prepared by Saturated Ammonium Sulfate (SAS) precipitation followed by dialysis.
- Pk:400 µl
Specifications
About this item
For WB starting dilution is: 1:1000. For IHC-P starting dilution is: 1:50~100.
Type: Primary
Antigen: CAMK2G
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 309-338
Host: Rabbit
Isotype: Ig
Reactivity: Human, Mouse