Anti-STK38 Rabbit Polyclonal Antibody
PRSI62-638
New Product
- Antibody type:Primary
- Antigen name:Serine/threonine-protein kinase 38
- Antigen symbol:STK38
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:410-439 aa
- Form:Liquid
- Gene ID:11329
- Modification:None
- Molecular weight:54 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:2 mg/ml
- Shipping temperature:4 °C
- Immunogen:This STK38 antibody is generated from rabbits immunised with a KLH conjugated synthetic peptide between 410-439 amino acids from the C-terminal region of human STK38.
- Purification:This antibody is prepared by Saturated Ammonium Sulfate (SAS) precipitation followed by dialysis.
- Pk:400 µl
For WB starting dilution is: 1:1000.
Type: Primary
Antigen: STK38
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 410-439
Host: Rabbit
Isotype: Ig
Reactivity: Human