Anti-GRK5 Rabbit Polyclonal Antibody
Catalog # PRSI62-585
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:G protein-coupled receptor kinase 5
- Antigen symbol:GRK5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:559-590 aa
- Form:Liquid
- Gene ID:2869
- Modification:None
- Molecular weight:68 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Shipping temperature:4 °C
- Immunogen:This GRK5 antibody is generated from rabbits immunised with a KLH conjugated synthetic peptide between 559-590 amino acids from the C-terminal region of human GRK5.
- Purification:This antibody is purified through a protein G column, followed by dialysis against PBS.
- Pk:400 µl
Specifications
About this item
For WB starting dilution is: 1:2000. For IHC-P starting dilution is: 1:50~100. For IHC-P starting dilution is: 1:50~100.
Type: Primary
Antigen: GRK5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 559-590
Host: Rabbit
Isotype: Ig
Reactivity: Human, Mouse