Anti-ORM1 Rabbit Polyclonal Antibody
Catalog # PRSI62-310
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:Alpha-1-acid glycoprotein 1
- Antigen symbol:ORM1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:152-180 aa
- Form:Liquid
- Gene ID:5004
- Modification:None
- Molecular weight:24 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:2 mg/ml
- Shipping temperature:4 °C
- Immunogen:This ORM1 antibody is generated from rabbits immunised with a KLH conjugated synthetic peptide between 152-180 amino acids from the C-terminal region of human ORM1.
- Purification:This antibody is prepared by Saturated Ammonium Sulfate (SAS) precipitation followed by dialysis.
- Pk:400 µl
Specifications
About this item
For WB starting dilution is: 1:1000. For FACS starting dilution is: 1:10~50.
Type: Primary
Antigen: ORM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 152-180
Host: Rabbit
Isotype: Ig
Reactivity: Human