Anti-CNBP Rabbit Polyclonal Antibody
Catalog # PRSI61-662
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:Cellular nucleic acid-binding protein
- Antigen symbol:CNBP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:67-96 aa
- Form:Liquid
- Gene ID:7555
- Modification:None
- Molecular weight:19 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,5 mg/ml
- Shipping temperature:4 °C
- Immunogen:This ZNF9 antibody is generated from rabbits immunised with a KLH conjugated synthetic peptide between 67-96 amino acids from the central region of human ZNF9.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Pk:400 µl
Specifications
About this item
For WB starting dilution is: 1:1000. For IHC-P starting dilution is: 1:50~100.
Type: Primary
Antigen: CNBP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 67-96
Host: Rabbit
Isotype: Ig
Reactivity: Human