Anti-MEP1A Rabbit Polyclonal Antibody
Catalog # PRSI23-421
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:Meprin A subunit alpha
- Antigen symbol:MEP1A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:4224
- Antigen synonyms:PPH alpha|N-benzoyl-L-tyrosyl-P-amino-benzoic acid hydrolase subunit alpha|Endopeptidase-2|MEP1A|Meprin A subunit alpha|PABA peptide hydrolase
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 440-700 of human MEP1A (NP_005579.2)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: MEP1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat