Anti-KDM5A Rabbit Polyclonal Antibody
PRSI22-804
New Product
- Antibody type:Primary
- Antigen name:Lysine demethylase 5A
- Antigen symbol:KDM5A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:5927
- Antigen synonyms:jumonji/ARID domain-containing protein 1A|Jumonji|RBP2|RBBP2|AT rich interactive domain 1A (RBP2-like)|histone demethylase JARID1A|lysine (K)-specific demethylase 5A|lysine-specific demethylase 5A|RBBP-2|retinoblastoma binding protein 2
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant protein of human KDM5A
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: KDM5A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat