Anti-NAP1L3 Rabbit Polyclonal Antibody
PRSI22-779
New Product
- Antibody type:Primary
- Antigen name:Nucleosome assembly protein 1-like 3
- Antigen symbol:NAP1L3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:4675
- Antigen synonyms:NAP1L3|BNAP|Nucleosome assembly protein 1-like 3
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 307-506 of human NAP1L3 (NP_004529.2)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: NAP1L3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human