Anti-MCM10 Rabbit Polyclonal Antibody
PRSI22-776
New Product
- Antibody type:Primary
- Antigen name:Minichromosome maintenance 10 replication initiation factor
- Antigen symbol:MCM10
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:55388
- Antigen synonyms:PRO2249|DNA43|MCM10 minichromosome maintenance deficient 1homolog of yeast MCM1hsMCM1minichromosome maintenance complex component 10|protein MCM10 homolog|CNA43
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 616-875 of human MCM10 (NP_877428.1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: MCM10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat