Anti-KIAA1456 Rabbit Polyclonal Antibody
PRSI22-772
New Product
- Antibody type:Primary
- Antigen name:TRNA methyltransferase 9B (putative)
- Antigen symbol:KIAA1456
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:57604
- Antigen synonyms:probable tRNA methyltransferase 9B|TRM9L|hTRM9L|KIAA1456|probable tRNA methyltransferase 9-like protein|C8orf79
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 110-330 of human KIAA1456 (NP_065895.2)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: KIAA1456
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat