Anti-COPS3 Rabbit Polyclonal Antibody
Catalog # PRSI22-634
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:COP9 signalosome subunit 3
- Antigen symbol:COPS3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:8533
- Antigen synonyms:COP9 constitutive photomorphogenic homolog subunit 3 (Arabidopsis)|CSN3|JAB1-containing signalosome subunit 3|COP9 constitutive photomorphogenic homolog subunit 3|SGN3|COPS3|COP9 complex subunit 3
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 194-423 of human COPS3 (NP_003644.2)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: COPS3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat