Anti-NELFE Rabbit Polyclonal Antibody
PRSI22-626
New Product
- Antibody type:Primary
- Antigen name:Negative elongation factor complex member E
- Antigen symbol:NELFE
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:7936
- Antigen synonyms:RD RNA-binding protein|D6S45|negative elongation factor E|nuclear protein|RDP|major histocompatibility complex gene RD|negative elongation factor polypeptide E|NELF-E|RDBP|RD|RNA-binding protein RD
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-280 of human NELFE (NP_002895.3)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: NELFE
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human