Anti-PARN Rabbit Polyclonal Antibody
PRSI22-571
New Product
- Antibody type:Primary
- Antigen name:Poly(A)-specific ribonuclease
- Antigen symbol:PARN
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:5073
- Antigen synonyms:DAN|poly(A)-specific ribonuclease PARN|polyadenylate-specific ribonuclease|PFBMFT4|deadenylating nuclease|DKCB6|deadenylation nuclease
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-280 of human PARN (NP_002573.1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: PARN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human