Anti-MYH1 Rabbit Polyclonal Antibody
Catalog # PRSI22-566
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:Myosin heavy chain 1
- Antigen symbol:MYH1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:4619
- Antigen synonyms:myosin-1|skeletal muscle|adult|epididymis luminal protein 71|adult 1|myosin heavy chain|myHC-IIx/d|MYHa|HEL71|myosin heavy chain 2x|heavy polypeptide 1|MYHSA1|myosin heavy chain IIx/d|myosin|MyHC-2x|heavy chain 1|MyHC-2X/D
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-100 of human MYH1 (NP_005954.3)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: MYH1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat