Anti-BLOC1S1 Rabbit Polyclonal Antibody
PRSI22-549
New Product
- Antibody type:Primary
- Antigen name:Biogenesis of lysosomal organelles complex 1 subunit 1
- Antigen symbol:BLOC1S1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:2647
- Antigen synonyms:GCN5-like protein 1|BLOS1|GCN5 (general control of amino-acid synthesis|biogenesis of lysosome-related organelles complex 1 subunit 1|MTA1-interacting coactivator|yeast|MICoA|BORCS1|GCN5 general control of amino-acid synthesis 5-like 1|RT14|GCN5L1|BLOC-1 subunit 1|homolog)-like 1
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 29-153 of human BLOC1S1 (NP_001478.2)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: BLOC1S1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse