Anti-RDM1 Rabbit Polyclonal Antibody
PRSI22-507
New Product
- Antibody type:Primary
- Antigen name:RAD52 motif containing 1
- Antigen symbol:RDM1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:201299
- Antigen synonyms:RAD52 motif-containing protein 1|RAD52 motif 1|RAD52B|RAD52 homolog B
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 125-284 of human RDM1 (NP_663629.1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: RDM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse